R&D Systems
Manufacturer Code:MAB17371SP
Catalog # MAB17371SP
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 805531 |
Immunogen | Mouse myeloma cell line NS0-derived recombinant human MIS/AMH Leu19-Gln450 (predicted) Accession # P03971 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: AMH; AMH MIF MIS Muellerian hormone muellerian-inhibiting factor muellerian-inhibiting substance Mullerian hormone Mullerian inhibiting factor Mullerian inhibiting substance; Anti-Muellerian hormone; MIS; Muellerian-inhibiting factor; Muellerian-inhibiting substance; Mullerian inhibiting factor; Mullerian inhibiting substance
Gene Aliases: AMH; MIF; MIS
UniProt ID: (Human) O75246
Entrez Gene ID: (Human) 268
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.