Novus Biologicals
Manufacturer Code:NBP290330V
Catalog # NB072355
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | 3 |
Immunogen | This antibody was obtained from a rabbit immunized with purified recombinant Human MIS/AMH (NP_000470.2 Leu25-Arg560). |
Conjugate | DyLight 405 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Rabbit Monoclonal Antibody
For Research Use Only
Protein Aliases: AMH; Anti-Muellerian hormone; MIF MIS Muellerian hormone muellerian-inhibiting factor muellerian-inhibiting substance Mullerian hormone Mullerian inhibiting factor Mullerian inhibiting substance; MIS; Muellerian-inhibiting factor; Muellerian-inhibiting substance; Mullerian inhibiting factor; Mullerian inhibiting substance
Gene Aliases: AMH; MIF; MIS
UniProt ID: (Human) O75246
Entrez Gene ID: (Human) 268
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.