Proteintech
Manufacturer Code:179771AP150UL
Catalog # 50-173-1913
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | MIP Fusion Protein Ag12369 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: AQP0 Aquaporin 0 LIM1 MIP MIP26 MP26; aquaporin 0; Aquaporin-0; dominant cataract; hydropic fibers; Lens fiber major intrinsic protein; lens opacity; major intrinsic protein of eye lens fiber; MIP26; MP26
Gene Aliases: AQP0; Cat; CTRCT15; Hfi; LIM1; Lop; MIP; MIP26; MP26; Palm; shrivelled; Svl
UniProt ID: (Human) P30301, (Mouse) P51180
Entrez Gene ID: (Human) 4284, (Mouse) 17339, (Rat) 25480
Molecular Function: transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.