Invitrogen
Manufacturer Code:MA523845
Catalog # PIMA523845
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Neutralization (Neu) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rat |
Class | Monoclonal |
Type | Antibody |
Clone | 87102 |
Immunogen | E. coli-derived recombit mouse CCL19/MIP-3 beta Gly26-Val107-Leu-Glu (Ser108LeuGlu) |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rat Monoclonal Antibody
For Research Use Only
Protein Aliases: C-C motif chemokine 19; chemokine CCL19; EBI-1 ligand chemokine; EBI1 ligand chemokine; EBI1-ligand chemokine; EBV-induced molecule 1 ligand chemokine; Epstein-Barr virus-induced molecule 1 ligand chemokine; small inducible cytokine A19; Small inducible cytokine A19 Macrophage inflammatory protein 3 beta MIP-3-beta EBI1-ligand chemokine ELC Beta chemokine exodus-3 CK beta-11; Small-inducible cytokine A19
Gene Aliases: Ccl19; CKb11; ELC; exodus-3; MIP3B; Scya19
UniProt ID: (Mouse) O70460
Entrez Gene ID: (Mouse) 24047
Molecular Function: chemokine cytokine signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.