Invitrogen
Manufacturer Code:PA546940
Catalog # PIPA546940
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Neutralization (Neu) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | E. coli-derived recombit human CCL19/MIP-3 beta Gly22-Ser98 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: beta chemokine exodus-3; Beta-chemokine exodus-3; C-C motif chemokine 19; CC chemokine ligand 19; chemokine (C-C motif) ligand 19; CK beta-11; EBI1 ligand chemokine; EBI1-ligand chemokine; Epstein-Barr virus-induced molecule 1 ligand chemokine; exodus-3; Macrophage inflammatory protein 3 beta; macrophage inflammatory protein 3-beta; MIP-3-beta; Small inducible cytokine A19 Macrophage inflammatory protein 3 beta MIP-3-beta EBI1-ligand chemokine ELC Beta chemokine exodus-3 CK beta-11; small inducible cytokine subfamily A (Cys-Cys), member 19; Small-inducible cytokine A19
Gene Aliases: CCL19; CKb11; ELC; MIP-3b; MIP3B; SCYA19
UniProt ID: (Human) Q99731
Entrez Gene ID: (Human) 6363
Molecular Function:
chemokine
cytokine
signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.