Invitrogen
Manufacturer Code:PA5114958
Catalog # PIPA5114958
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human CCL19 corresponding to a region within N-terminal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: beta chemokine exodus-3; beta chemokine exodus-3 beta-chemokine exodus-3 CC chemokine ligand 19 C-C motif chemokine C-C motif chemokine 19 C-C motif chemokine ligand 19 Ccl19 chemokine (C-C motif) ligand 19 chemokine CCL19 CK beta-11 CKb11 EBI1 ligand chemokine EBI-1 ligand chemokine EBI1-ligand chemokine EBV-induced molecule 1 ligand chemokine ELC Epstein-Barr virus-induced molecule 1 ligand chemokine exodus-3 Macrophage inflammatory protein 3 beta macrophage inflammatory protein 3-beta MIP3B MIP-3b MIP-3-beta M-MIP-3-beta Scya19 small inducible cytokine A19 small inducible cytokine subfamily A (Cys-Cys) member 19 small-inducible cytokine A19; Beta-chemokine exodus-3; C-C motif chemokine 19; CC chemokine ligand 19; chemokine (C-C motif) ligand 19; chemokine CCL19; CK beta-11; EBI-1 ligand chemokine; EBI1 ligand chemokine; EBI1-ligand chemokine; EBV-induced molecule 1 ligand chemokine; Epstein-Barr virus-induced molecule 1 ligand chemokine; exodus-3; Macrophage inflammatory protein 3 beta; macrophage inflammatory protein 3-beta; MIP-3-beta; small inducible cytokine A19; small inducible cytokine subfamily A (Cys-Cys), member 19; Small-inducible cytokine A19
Gene Aliases: CCL19; CKb11; ELC; exodus-3; MIP-3b; MIP3B; SCYA19
UniProt ID: (Human) Q99731, (Mouse) O70460
Entrez Gene ID: (Human) 6363, (Mouse) 24047
Molecular Function:
chemokine
cytokine
signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.