Invitrogen
Manufacturer Code:MA523548
Catalog # PIMA523548
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rat |
Class | Monoclonal |
Type | Antibody |
Clone | 114906 |
Immunogen | E. coli-derived recombit mouse CCL20/MIP-3 alpha Ala28-Met97 |
Conjugate | FITC |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | 4° C store in dark DO NOT FREEZE! |
Rat Monoclonal Antibody
For Research Use Only
Protein Aliases: beta chemokine exodus-1; Beta-chemokine exodus-1; C-C motif chemokine 20; CC chemokine LARC; CC chemokine ST38; CKb4 LARC MIP-3a MIP3A SCYA20 ST38 C-C motif chemokine 20 CC chemokine LARC MIP-3-alpha beta chemokine exodus-1 beta-chemokine exodus-1 exodus-1 liver and activation-regulated chemokine macrophage inflammatory protein 3 alpha small inducible cytokine subfamily A (Cys-Cys) member 20 small-inducible cytokine A20; Liver and activation-regulated chemokine; Macrophage inflammatory protein 3 alpha; MIP-3-alpha; small inducible cytokine subfamily A20; Small-inducible cytokine A20
Gene Aliases: Ccl20; CKb4; exodus-1; LARC; MIP-3[a]; MIP-3A; MIP3A; Scya20; ST38
UniProt ID: (Mouse) O89093
Entrez Gene ID: (Mouse) 20297
Molecular Function: chemokine cytokine signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.