Invitrogen
Manufacturer Code:PA530650
Catalog # PIPA530650
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein fragment corresponding to a region within amino acids 1 and 204 of Human myo-inositol oxygenase |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: aldehyde reductase (aldose reductase) like 6; aldehyde reductase (aldose reductase)-like 6; aldehyde reductase 6, renal; Aldehyde reductase-like 6; ALDRL6 MI oxygenase aldehyde reductase (aldose reductase) like 6 aldehyde reductase-like 6 inositol oxygenase kidney-specific protein 32 renal-specific oxidoreductase; Inositol oxygenase; Kidney-specific protein 32; MI oxygenase; Myo-inositol oxygenase; renal-specific oxido-reducatse; renal-specific oxido-reductase; Renal-specific oxidoreductase
Gene Aliases: 0610009I10Rik; AI314022; ALDRL6; C85427; KSP32; MIOX; RSOR
UniProt ID: (Human) Q9UGB7, (Rat) Q9QXN4, (Mouse) Q9QXN5
Entrez Gene ID: (Human) 55586, (Rat) 252899, (Mouse) 56727
Molecular Function:
oxidoreductase
oxygenase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.