Proteintech
Manufacturer Code:208261AP150UL
Catalog # 50-173-3110
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | MIOS Fusion Protein Ag14802 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: GATOR complex protein MIOS; H_DJ1159O04.1; MIOS; Missing oocyte meiosis regulator homolog; missing oocyte, meiosis regulator, homolog; WD repeat-containing protein mio; WUGSC:H_DJ1159O04.1
Gene Aliases: MIO; MIOS; RGD1308432; Sea4
UniProt ID: (Human) Q9NXC5
Entrez Gene ID: (Human) 54468, (Rat) 362324
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.