OriGene
Manufacturer Code:TA507234
Catalog # 50-167-7019
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI1B2 |
Immunogen | Full length human recombit protein of human MINPP1 produced in HEK293T cell. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: 2,3-bisphosphoglycerate 3-phosphatase; 2,3-BPG phosphatase; HIPER1 MINPP2 MIPP; Inositol (1,3,4,5)-tetrakisphosphate 3-phosphatase; Ins(1,3,4,5)P(4) 3-phosphatase; multiple inositol polyphosphate histidine phosphatase, 1; Multiple inositol polyphosphate phosphatase 1; multiple inositol polyphosphate phosphatase 2
Gene Aliases: HIPER1; MINPP1; MINPP2; MIPP; UNQ900/PRO1917
UniProt ID: (Human) Q9UNW1
Entrez Gene ID: (Human) 9562
Molecular Function: hydrolase nucleotide phosphatase phosphatase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.