Proteintech
Manufacturer Code:122141AP150UL
Catalog # 50-172-7800
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | MINA Fusion Protein Ag2838 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 60S ribosomal protein L27a histidine hydroxylase; Bifunctional lysine-specific demethylase and histidyl-hydroxylase MINA; Histone lysine demethylase MINA; MDIG MINA Mina53 MYC induced nuclear antigen NO52 Nucleolar protein 52; MINA; mina {ECO:0000312|RGD:708521}; Mina-53; mineral dust induced gene protein; Mineral dust-induced gene protein; MYC-induced nuclear antigen; myc-induced nuclear antigen, 53 kDa; Nucleolar protein 52; Ribosomal oxygenase 2; Ribosomal oxygenase MINA; ROX
Gene Aliases: 1810047J07Rik; 2410057H13Rik; 3830408E23Rik; AI449204; MDIG; MINA; MINA53; NO52; RIOX2; ROX
UniProt ID: (Human) Q8IUF8, (Mouse) Q8CD15, (Rat) Q8CFC1
Entrez Gene ID: (Human) 84864, (Mouse) 67014, (Rat) 266670
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.