R&D Systems
Manufacturer Code:MAB19781
Catalog # MAB19781
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Western Blot (WB) | Assay dependent |
CyTOF (CyTOF) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rat |
Class | Monoclonal |
Type | Antibody |
Clone | 811409 |
Immunogen | <EM>E. coli</EM>-derived recombinant mouse MIF Met1-Ala115 Accession # P34884 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Rat Monoclonal Antibody
Protein Aliases: EC 5.3.2.1 EC 5.3.3.12 GIFmacrophage migration inhibitory factor GLIF Glycosylation-inhibiting factor L-dopachrome isomerase L-dopachrome tautomerase macrophage migration inhibitory factor (glycosylation-inhibiting factor) MMIF Phenylpyruvate tautomerase; GIF; Glycosylation-inhibiting factor; L-dopachrome isomerase; L-dopachrome tautomerase; Macrophage migration inhibitory factor; MIF; Phenylpyruvate tautomerase
Gene Aliases: GIF; GLIF; MIF; MMIF
UniProt ID: (Human) P14174
Entrez Gene ID: (Human) 4282
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.