Proteintech
Manufacturer Code:204151AP150UL
Catalog # 50-173-2876
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | MIF Fusion Protein Ag14058 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Delayed early response protein 6; DER6; GIF; GIF GLIF L dopachrome isomerase L dopachrome tautomerase MIF MMIF Phenylpyruvate tautomerase; Glutathione-binding 13 kDa protein; Glycosylation-inhibiting factor; L-dopachrome isomerase; L-dopachrome tautomerase; Macrophage migration inhibitory factor; MIF; Phenylpyruvate tautomerase
Gene Aliases: GIF; GLIF; MIF; MMIF
UniProt ID: (Human) P14174, (Mouse) P34884, (Rat) P30904
Entrez Gene ID: (Human) 4282, (Mouse) 17319, (Rat) 81683
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.