Invitrogen
Manufacturer Code:PA540896
Catalog # PIPA540896
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the middle region of human MIER1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Early response 1; mesoderm induction early response 1 homolog; Mesoderm induction early response 1 homolog (Xenopus laevis) ER1 MI-ER1 hMI-ER1 RP5-944N15.1; mesoderm induction early response 1, transcriptional regulator; Mesoderm induction early response protein 1
Gene Aliases: 4933425I22Rik; 5830411K19Rik; ER1; KIAA1610; MI-ER1; MIER1; RGD1562337
UniProt ID: (Human) Q8N108, (Mouse) Q5UAK0
Entrez Gene ID: (Rabbit) 100354543, (Human) 57708, (Horse) 100053862, (Dog) 479536, (Bovine) 538742, (Guinea pig) 100723878, (Mouse) 71148, (Rat) 313418
Molecular Function: DNA binding protein chromatin/chromatin-binding protein histone nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.