Novus Biologicals
Manufacturer Code:NBP181730
Catalog # NBP181730
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | This antibody was developed against Recombinant Protein corresponding to amino acids:MMEGETNFSSEIEDLAREGDMPIHELLSLYGYGSTVRLPEEDEEEEEEEEEGEDDEDADNDDNSGCSGENKEENIKDSSGQEDETQSSNDDPSQSVASQDAQEI |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: DKFZp781G0451 Early response 1 ER1 hMI-ER1 KIAA1610MI-ER1 mesoderm induction early response 1 (MI-ER1) mesoderm induction early response 1 homolog (Xenopus laevis) mesoderm induction early response protein 1 MGC131940 MGC150640 MGC150641 Mi-er1; Early response 1; mesoderm induction early response 1 homolog; mesoderm induction early response 1, transcriptional regulator; Mesoderm induction early response protein 1
Gene Aliases: ER1; KIAA1610; MI-ER1; MIER1
UniProt ID: (Human) Q8N108
Entrez Gene ID: (Human) 57708
Molecular Function:
DNA binding protein
chromatin/chromatin-binding protein
histone
nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.