Abnova
Manufacturer Code:H00057708K
Catalog # 89-210-533
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Immunogen | A synthetic peptide of human MIER1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Rabbit monoclonal antibody raised against a human MIER1 peptide using ARM Technology.
For Research Use Only
Protein Aliases: DKFZp781G0451 ER1 KIAA1610 MGC131940 MGC150640 MGC150641 MI-ER1 RP5-944N15.1 hMI-ER1; Early response 1; mesoderm induction early response 1 homolog; mesoderm induction early response 1, transcriptional regulator; Mesoderm induction early response protein 1
Gene Aliases: ER1; KIAA1610; MI-ER1; MIER1
UniProt ID: (Human) Q8N108
Entrez Gene ID: (Human) 57708
Molecular Function: DNA binding protein chromatin/chromatin-binding protein histone nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.