Invitrogen
Manufacturer Code:PA545891
Catalog # PIPA545891
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the C-terminal of human MICU1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ara CALC; Atopy-related autoantigen CALC; calcium binding atopy-related autoantigen 1; Calcium uptake protein 1, mitochondrial; calcium-binding atopy-like autoantigen 1; Calcium-binding atopy-related autoantigen 1; Calcium-binding atopy-related autoantigen 1 homolog; calcium-binding atopy-related autoantigen-like protein; Hom s 4; mitochondrial Calcium uptake 1 CALC DKFZp564C246 EFHA3 FLJ12684 CBARA1
Gene Aliases: C730016L05Rik; CALC; CBARA1; EFHA3; MICU1; MPXPS; si:ch211-199i24.4; zgc:162148
UniProt ID: (Bovine) Q0IIL1, (Human) Q9BPX6, (Mouse) Q8VCX5, (Rat) Q6P6Q9, (Zebrafish) A4IG32
Entrez Gene ID: (Dog) 479241, (Goat) 100861082, (Bovine) 507899, (Horse) 100063651, (Rabbit) 100341636, (Human) 10367, (Mouse) 216001, (Guinea pig) 100715334, (Rat) 365567, (Zebrafish) 561210
Molecular Function:
calcium-binding protein
calmodulin
intracellular calcium-sensing protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.