R&D Systems
Manufacturer Code:FAB1599N025
Catalog # FAB1599N025
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 236511 |
Immunogen | Mouse myeloma cell line NS0-derived recombinant human MICB Ala23-Gly298 Accession # CAI18747 |
Conjugate | Alexa Fluor 700 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Protect from light. Do not freeze. 12 months from date of receipt 2 to 8 degreesC as supplied. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: MHC class I chain-related protein B; MHC class I chain-related protein B MHC class I mic-B antigen MHC class I polypeptide-related sequence B MIC-B PERB11.2MHC class I-like molecule PERB11.2-IMX stress inducible class I homolog; MHC class I mic-B antigen; MHC class I polypeptide-related sequence B; MHC class I-like molecule PERB11.2-IMX; MIC-B; MICB; stress inducible class I homolog
Gene Aliases: MICB; PERB11.2
UniProt ID: (Human) Q29980
Entrez Gene ID: (Human) 4277
Molecular Function: cytokine receptor defense/immunity protein immunoglobulin receptor superfamily major histocompatibility complex antigen receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.