Invitrogen
Manufacturer Code:PA579672
Catalog # PIPA579672
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to a sequence at the N-terminus of human MICB. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: MHC class I chain-related protein B; MHC class I mic-B antigen; MHC class I polypeptide-related sequence B; MHC class I polypeptide-related sequence B MIC-B MICB; MHC class I-like located near the leukocyte receptor complex 2; MHC class I-like located near the LRC, 2; MHC class I-like molecule PERB11.2-IMX; MHC class I-like protein MILL2; MHC I like leukocyte 2; MIC-B; MICB; Mill2 gene for MHC class I-like located near the LRC, 2, exon 3, partial cds, strain:LEW.1lm1; stress inducible class I homolog
Gene Aliases: MICB; Mill2; PERB11.2
UniProt ID: (Human) Q29980, (Rat) A0A0G2K7V7
Entrez Gene ID: (Human) 4277, (Rat) 365212
Molecular Function: cytokine receptor defense/immunity protein immunoglobulin receptor superfamily major histocompatibility complex antigen receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.