Abnova
Manufacturer Code:H00004277B01
Catalog # 89-005-765
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Polyclonal |
Type | Antibody |
Immunogen | MICB (AAH44218.1 1 a.a. ~ 340 a.a) full-length human protein. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Mouse polyclonal antibody raised against a full-length human MICB protein.
For Research Use Only
Protein Aliases: MHC class I chain-related protein B; MHC class I mic-B antigen; MHC class I polypeptide-related sequence B; MHC class I-like molecule PERB11.2-IMX; MIC-B; MICB; PERB11.2; stress inducible class I homolog
Gene Aliases: MICB; PERB11.2
UniProt ID: (Human) Q29980
Entrez Gene ID: (Human) 4277
Molecular Function:
cytokine receptor
defense/immunity protein
immunoglobulin receptor superfamily
major histocompatibility complex antigen
receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.