Invitrogen
Manufacturer Code:PA5100613
Catalog # PIPA5100613
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human MICAL2 corresponding to a region within the internal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: [F-actin]-monooxygenase MICAL2; [F-actin]-monooxygenase MICAL2 5330438E18Rik 9530064J02 flavoprotein oxidoreductase MICAL2 KIAA0750 Mical2 MICAL-2 MICAL2PV1 MICAL2PV2 microtubule associated monooxygenase calponin and LIM domain containing 2 microtubule associated monoxygenase calponin and LIM domain containing 2 mKIAA0750 mMical2 molecule interacting with CasL protein 2 protein MICAL-2 protein-methionine sulfoxide oxidase MICAL2 RGD1311773; flavoprotein oxidoreductase MICAL2; MICAL-2; microtubule associated monoxygenase, calponin and LIM domain containing 2; mMical2; Molecule interacting with CasL protein 2; protein MICAL-2; protein-methionine sulfoxide oxidase MICAL2
Gene Aliases: 5330438E18Rik; 9530064J02; KIAA0750; MICAL-2; MICAL2; MICAL2PV1; MICAL2PV2; mKIAA0750; RGD1311773
UniProt ID: (Human) O94851, (Rat) D4A1F2, (Mouse) Q8BML1
Entrez Gene ID: (Human) 9645, (Rat) 365352, (Mouse) 320878
Molecular Function:
oxidoreductase
oxygenase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.