Abnova
Manufacturer Code:PAB22448
Catalog # 89-124-156
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (PFA fixed) (IHC (PFX)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant protein corresponding to amino acids of human MICAL2. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C. For long term storage store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against recombinant MICAL2.
For Research Use Only
Protein Aliases: [F-actin]-monooxygenase MICAL2; DKFZp686H03148; DKFZp686H2469; flavoprotein oxidoreductase MICAL2; KIAA0750; MICAL-2; MICAL2PV1; MICAL2PV2; microtubule associated monoxygenase, calponin and LIM domain containing 2; Molecule interacting with CasL protein 2
Gene Aliases: KIAA0750; MICAL-2; MICAL2; MICAL2PV1; MICAL2PV2
UniProt ID: (Human) O94851
Entrez Gene ID: (Human) 9645
Molecular Function:
oxidoreductase
oxygenase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.