Abnova
Manufacturer Code:PAB15772
Catalog # 89-117-836
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant GST fusion protein corresponding to 168 amino acids of mouse Mical2. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against partial recombinant Mical2.
For Research Use Only
Protein Aliases: 5330438E18Rik; 9530064J02; [F-actin]-monooxygenase MICAL2; KIAA0750; MICAL-2; microtubule associated monoxygenase, calponin and LIM domain containing 2; mKIAA0750; mMical2; Molecule interacting with CasL protein 2; protein MICAL-2; protein-methionine sulfoxide oxidase MICAL2
Gene Aliases: 5330438E18Rik; 9530064J02; Kiaa0750; MICAL-2; Mical2; mKIAA0750
UniProt ID: (Mouse) Q8BML1
Entrez Gene ID: (Mouse) 320878
Molecular Function:
oxidoreductase
oxygenase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.