Invitrogen
Manufacturer Code:PA5106316
Catalog # PIPA5106316
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human MICAL1 corresponding to a region within the internal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: [F-actin]-monooxygenase MICAL1; [F-actin]-monooxygenase MICAL1 CasL interacting molecule MICAL MICAL MICAL1 MICAL-1 microtubule associated monooxygenase calponin and LIM domain containing 1 microtubule associated monoxygenase calponin and LIM domain containing 1 mMical1 molecule interacting with CasL protein 1 NEDD9 interacting protein with calponin homology and LIM domains NEDD9-interacting protein with calponin homology and LIM domains NICAL protein-methionine sulfoxide oxidase MICAL1; CasL interacting molecule MICAL; MICAL-1; microtubule associated monoxygenase, calponin and LIM domain containing 1; mMical1; Molecule interacting with CasL protein 1; NEDD9 interacting protein with calponin homology and LIM domains; NEDD9-interacting protein with calponin homology and LIM domains; protein-methionine sulfoxide oxidase MICAL1
Gene Aliases: MICAL; MICAL-1; MICAL1; NICAL
UniProt ID: (Human) Q8TDZ2, (Mouse) Q8VDP3
Entrez Gene ID: (Human) 64780, (Mouse) 171580
Molecular Function:
actin family cytoskeletal protein
cytoskeletal protein
non-motor actin binding protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.