Invitrogen
Manufacturer Code:PA528181
Catalog # PIPA528181
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein fragment corresponding to a region within amino acids 33 and 276 of Human MICA |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: HLA class I antigen; MHC class I chain-related protein A; MHC class I polypeptide-related sequence A; MHC class I related sequence A; MIC-A; MICA; stress inducible class I homolog; truncated MHC class I polypeptide-related sequence A
Gene Aliases: MIC-A; MICA; PERB11.1
UniProt ID: (Human) Q29983
Entrez Gene ID: (Human) 100507436
Molecular Function: cytokine receptor defense/immunity protein immunoglobulin receptor superfamily major histocompatibility complex antigen receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.