R&D Systems
Manufacturer Code:BAF1300
Catalog # BAF1300
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | Mouse myeloma cell line NS0-derived recombinant human MICA Ala22-Gln308 Accession # NP_001170990 |
Conjugate | Biotin |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: FLJ60820 MGC111087 PERB11.1; HLA class I antigen; MHC class I chain-related protein A; MHC class I polypeptide-related sequence A; MHC class I related sequence A; MIC-A; MICA; stress inducible class I homolog; truncated MHC class I polypeptide-related sequence A
Gene Aliases: MIC-A; MICA; PERB11.1
UniProt ID: (Human) Q29983
Entrez Gene ID: (Human) 100507436
Molecular Function: cytokine receptor defense/immunity protein immunoglobulin receptor superfamily major histocompatibility complex antigen receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.