Invitrogen
Manufacturer Code:17532181
Catalog # 50-112-9473
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rat |
Class | Monoclonal |
Type | Antibody |
Clone | M5/114.15.2 |
Conjugate | APC |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | 4° C store in dark DO NOT FREEZE! |
Rat Monoclonal Antibody
For Research Use Only
Protein Aliases: cell surface glycoprotein; H-2 class II histocompatibility antigen, A beta chain; H-2 class II histocompatibility antigen, I-E beta chain; histocompatibility 2, class II antigen A, beta 1; I-Ab I-Ad I-Aq I-Ed I-Ek MHC class 2 MHC class II glygoproteins; I-E beta MHC class II; major histocompatibility complex class II beta chain; major histocompatibility complex H-2E beta chain; MHC class II antigen A beta; MHC class II H2-IA-beta-psi; MHC class II IE-b; MHC H2-IE-beta cell surface glycoprotein; response to metastatic cancers 1
Gene Aliases: Abeta; AI845868; H-2Ab; H-2Eb; H2-Ab; H2-Ab1; H2-iabeta; H2Eb; I-Abeta; Ia-2; Ia-4; Ia2; Ia4; IAb; Rmcs1
UniProt ID: (Mouse) P14483
Entrez Gene ID: (Mouse) 14961, (Mouse) 14969
Molecular Function:
defense/immunity protein
major histocompatibility complex antigen
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.