Novus Biologicals
Manufacturer Code:NB1206405
Catalog # NB1206405
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Frozen) (IHC (F)) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Neutralization (Neu) | Assay dependent |
CyTOF (CyTOF) | Assay dependent |
Species reactivity | Rat |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OX18 |
Immunogen | Rat spleen glycoproteins |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: Clone OX18 DKFZp686P19218 EA1.2 EA2.1 HLA class I histocompatibility antigen alpha chain E HLA class I histocompatibility antigen E alpha chain HLA-6.2 HLAE lymphocyte antigen major histocompatibility complex class I E MHC MHC class I antigen E MHC Class I RT1-A MHC HLA-E alpha-1 MHC HLA-E alpha-2.1 OX18 QA1 RT1-A RT1-A (OX18); HLA class I histocompatibility antigen, alpha chain E; HLA class I histocompatibility antigen, E alpha chain; lymphocyte antigen; MHC class I antigen E; MHC HLA-E alpha-1; MHC HLA-E alpha-2.1; sHLA-E; Soluble HLA class I histocompatibility antigen, alpha chain E
Gene Aliases: EA1.2; EA2.1; HLA-6.2; HLA-E; HLAE; MHC; QA1
UniProt ID: (Human) P13747
Entrez Gene ID: (Human) 3133
Molecular Function: cytokine receptor defense/immunity protein immunoglobulin receptor superfamily major histocompatibility complex antigen receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.