Novus Biologicals
Manufacturer Code:NB500307PE
Catalog # NB500307PE
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | MEM-E/06 |
Immunogen | Bacterially expressed recombinant full length protein refolded with beta2-microglobulin and peptide (Human). |
Conjugate | PE |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C in the dark. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: DKFZp686P19218 EA1.2 EA2.1 HLA class I histocompatibility antigen alpha chain E HLA class I histocompatibility antigen E alpha chain HLA-6.2 HLAE lymphocyte antigen major histocompatibility complex class I E MHC MHC class I antigen E MHC HLA-E alpha-1 MHC HLA-E alpha-2.1 QA1; HLA class I histocompatibility antigen, alpha chain E; HLA class I histocompatibility antigen, E alpha chain; lymphocyte antigen; MHC class I antigen E; MHC HLA-E alpha-1; MHC HLA-E alpha-2.1; sHLA-E; Soluble HLA class I histocompatibility antigen, alpha chain E
Gene Aliases: EA1.2; EA2.1; HLA-6.2; HLA-E; HLAE; MHC; QA1
UniProt ID: (Human) P13747
Entrez Gene ID: (Human) 3133
Molecular Function: cytokine receptor defense/immunity protein immunoglobulin receptor superfamily major histocompatibility complex antigen receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.