Abnova
Manufacturer Code:H00004258M01A
Catalog # 89-104-927
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 1D11 |
Immunogen | MGST2 (AAH254161 a.a. &bsim 147 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Mouse monoclonal antibody raised against a full-length recombinant MGST2.
For Research Use Only
Protein Aliases: FLJ27438; Glutathione peroxidase MGST2; GST2; Leukotriene C4 synthase MGST2; MGC14097; MGST-II; Microsomal glutathione S-transferase 2; Microsomal glutathione S-transferase II; Microsomal GST-2; Microsomal GST-II
Gene Aliases: GST2; MGST-II; MGST2
UniProt ID: (Human) Q99735
Entrez Gene ID: (Human) 4258
Molecular Function:
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.