Invitrogen
Manufacturer Code:PA572428
Catalog # PIPA572428
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | region 1 - 60 aa |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Glutathione peroxidase MGST2; Leukotriene C4 synthase MGST2; Microsomal glutathione S-transferase 2; Microsomal glutathione S-transferase 2 FLJ27438 GST2 MGC14097 MGST-II; Microsomal glutathione S-transferase II; Microsomal GST-2; Microsomal GST-II
Gene Aliases: GST2; MGST-II; MGST2
UniProt ID: (Human) Q99735, (Mouse) A2RST1
Entrez Gene ID: (Human) 4258, (Rat) 295037, (Mouse) 211666
Molecular Function: transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.