Novus Biologicals
Manufacturer Code:NBP159066
Catalog # NBP159066
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptides corresponding to MGST2(microsomal glutathione S-transferase 2) The peptide sequence was selected from the N terminal of MGST2. Peptide sequence MAGNSILLAAVSILSACQQSYFALQVGKARLKYKVTPPAVTGSPEFERVF. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: EC 2.5.1.18 FLJ27438 GST2MGC14097 MGST-II microsomal glutathione S-transferase 2 Microsomal GST-2 Microsomal GST-II truncated microsomal glutathione S-transferase 2; Glutathione peroxidase MGST2; Leukotriene C4 synthase MGST2; Microsomal glutathione S-transferase 2; Microsomal glutathione S-transferase II; Microsomal GST-2; Microsomal GST-II
Gene Aliases: GST2; MGST-II; MGST2
UniProt ID: (Human) Q99735
Entrez Gene ID: (Human) 4258
Molecular Function: transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.