Novus Biologicals
Manufacturer Code:NBP272660IR
Catalog # NBP272660IR
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI11G6 |
Immunogen | Full length recombinant protein of human MGP (NP_000891) produced in E.coli. |
Conjugate | DyLight 755 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Mouse Monoclonal Antibody
Protein Aliases: Cell growth-inhibiting gene 36 protein; Cell growth-inhibiting gene 36 protein matrix Gla protein MGLAP NTI; Matrix Gla protein; MGP
Gene Aliases: GIG36; MGLAP; MGP; NTI
UniProt ID: (Human) P08493
Entrez Gene ID: (Human) 4256
Molecular Function:
annexin
calcium-binding protein
calmodulin
extracellular matrix protein
extracellular matrix structural protein
intracellular calcium-sensing protein
signaling molecule
structural protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.