Proteintech
Manufacturer Code:107341AP150UL
Catalog # 50-172-6550
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | MGP Fusion Protein Ag1091 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Cell growth-inhibiting gene 36 protein; GIG36 matrix Gla protein MGLAP MGP NTI; Matrix Gla protein; MGP
Gene Aliases: GIG36; MGLAP; MGP; NTI
UniProt ID: (Human) P08493
Entrez Gene ID: (Human) 4256
Molecular Function:
annexin
calcium-binding protein
calmodulin
extracellular matrix protein
extracellular matrix structural protein
intracellular calcium-sensing protein
signaling molecule
structural protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.