Novus Biologicals
Manufacturer Code:NB100168SS
Catalog # NB100168SS
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | MT 23.2 |
Immunogen | Recombinant human MGMT (O6-methylguanine-DNA methyltransferase) purified from E. coli [UniProt# P16455] |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: 6-O-methylguanine-DNA methyltransferase; 6-O-methylguanine-DNA methyltransferase EC 2.1.1.63 methylated-DNA--protein-cysteine methyltransferase methylguanine-DNA methyltransferase O-6-methylguanine-DNA methyltransferase O6-methylguanine-DNA methyltransferase O-6-methylguanine-DNA-alkyltransferase; methylated DNA protein cysteine methyltransferase; Methylated-DNA--protein-cysteine methyltransferase; methylguanine-DNA methyltransferase; MGMT; O-6-methylguanine-DNA-alkyltransferase; O6-methylguanine-DNA methyltransferase
Gene Aliases: MGMT
UniProt ID: (Human) P16455
Entrez Gene ID: (Human) 4255
Molecular Function: DNA binding protein DNA methyltransferase methyltransferase nucleic acid binding transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.