Invitrogen
Manufacturer Code:PA533822
Catalog # PIPA533822
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic 19 amino acid peptide from N-terminal extracellular domain of human GRM4 / MGLUR4. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: glutamate receptor, metabotropic 4; GRM4 GPRC1D MGlu4 MGLUR4; Metabotropic glutamate receptor 4; mGluR4
Gene Aliases: GPRC1D; GRM4; mGlu4; MGLUR4
UniProt ID: (Human) Q14833
Entrez Gene ID: (Human) 2914
Molecular Function:
G-protein coupled receptor
receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.