Proteintech
Manufacturer Code:149861AP150UL
Catalog # 50-172-9888
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | MGLL Fusion Protein Ag6970 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: HU K5 HUK5 Lysophospholipase homolog Lysophospholipase like MGL MGLL Monoglyceride lipase; HU-K5; Lysophospholipase homolog; Lysophospholipase-like; MAGL; MGL; Monoacylglycerol lipase; Monoglyceride lipase
Gene Aliases: AA589436; HU-K5; HUK5; MAGL; MGL; Mgl2; MGLL
UniProt ID: (Human) Q99685, (Rat) Q8R431, (Mouse) O35678
Entrez Gene ID: (Human) 11343, (Rat) 29254, (Mouse) 23945
Molecular Function:
hydrolase
lipase
phospholipase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.