Invitrogen
Manufacturer Code:PA535926
Catalog # PIPA535926
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit fragment corresponding to a region within amino acids 131 and 849 of MGEA5 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Beta-hexosaminidase; Beta-N-acetylhexosaminidase; Bifunctional protein NCOAT; Meningioma-expressed antigen 5; N-acetyl-beta-D-glucosaminidase; N-acetyl-beta-glucosaminidase; nuclear cytoplasmic O-GlcNAcase and acetyltransferase; OGA; OGA MEA5 NCOAT meningioma expressed antigen 5 hyaluronidase; Protein O-GlcNAcase
Gene Aliases: 2810009A20Rik; 4833427O07Rik; 5830447M11Rik; AA408215; Hexc; Hy5; Kiaa0679; Mea5; Mgea5; mKIAA0679; Ncoat; O-GLCNACASE; OGA
UniProt ID: (Rat) Q8VIJ5, (Mouse) Q9EQQ9
Entrez Gene ID: (Rat) 154968, (Mouse) 76055
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.