Proteintech
Manufacturer Code:169931AP150UL
Catalog # 50-173-1316
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | MGAT5B Fusion Protein Ag10678 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Alpha-1,6-mannosylglycoprotein 6-beta-N-acetylglucosaminyltransferase B; Alpha-mannoside beta-1,6-N-acetylglucosaminyltransferase B; beta(1,6)-N-acetylglucosaminyltransferase V; GlcNAc T Vb GnT IX GnT VB hGnTVb KIAA2008 MGAT5B; GlcNAc-T Vb; GNT-IX; GNT-Vb; hGnTVb; mannoside acetylglucosaminyltransferase 5, isoenzyme B; Mannoside acetylglucosaminyltransferase 5B; mannosyl (alpha-1,6-)-glycoprotein beta-1,6-N-acetyl-glucosaminyltransferase, isoenzyme B; N-acetylglucosaminyl-transferase Vb; N-acetylglucosaminyltranferase VB; N-acetylglucosaminyltransferase IX
Gene Aliases: GnT-IX; GnT-VB; KIAA2008; MGAT5B
UniProt ID: (Human) Q3V5L5
Entrez Gene ID: (Human) 146664, (Rat) 303693
Molecular Function: glycosyltransferase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.