Invitrogen
Manufacturer Code:PA5100611
Catalog # PIPA5100611
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human MGAT4C corresponding to a region within the internal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 9130411I17Rik Alpha-13-mannosyl-glycoprotein 4-beta-N-acetylglucosaminyltransferase C glcNAc-T IVc GnT-IVc GNTIVH HGNT-IV-H mannosyl (alpha-13-)-glycoprotein beta-14-N-acetylglucosaminyltransferase isozyme C mannosyl (alpha-13-)-glycoprotein beta-14-N-acetylglucosaminyltransferase isozyme C (putative) MGAT4 family member C MGAT4 family member C MGAT4C N-acetylglucosaminyltransferase IV homolog N-acetylglucosaminyltransferase IVc N-glycosyl-oligosaccharide-glycoprotein N-acetylglucosaminyltransferase IVc RGD1307871 UDP-N-acetylglucosamine: alpha-13-D-mannoside beta-14-N-acetylglucosaminyltransferase IVc UDP-N-acetylglucosamine:a-13-D-mannoside beta-14-N-acetylglucosaminyltransferase IV UDP-N-acetylglucosamine:a-13-D-mannoside beta-14-N-acetylglucosaminyltransferase IV-homolog; Alpha-1,3-mannosyl-glycoprotein 4-beta-N-acetylglucosaminyltransferase C; GlcNAc-T IVc; GnT-IVc; hGnT-IV-H; mannosyl (alpha-1,3-)-glycoprotein beta-1,4-N-acetylglucosaminyltransferase, isozyme C (putative); MGAT4 family, member C; N-acetylglucosaminyltransferase IV homolog; N-acetylglucosaminyltransferase IVc; N-glycosyl-oligosaccharide-glycoprotein N-acetylglucosaminyltransferase IVc; UDP-N-acetylglucosamine: alpha-1,3-D-mannoside beta-1,4-N-acetylglucosaminyltransferase IVc; UDP-N-acetylglucosamine:a-1,3-D-mannoside beta-1,4-N-acetylglucosaminyltransferase IV; UDP-N-acetylglucosamine:a-1,3-D-mannoside beta-1,4-N-acetylglucosaminyltransferase IV-homolog
Gene Aliases: 9130411I17Rik; GNTIVH; HGNT-IV-H; MGAT4C; RGD1307871
UniProt ID: (Human) Q9UBM8, (Mouse) Q9D306
Entrez Gene ID: (Human) 25834, (Rat) 299756, (Mouse) 67569
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.