Novus Biologicals
Manufacturer Code:NBP30493420UL
Catalog # NB096196
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 299-478 of human MGAT4C (NP_037376.2). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Alpha-1,3-mannosyl-glycoprotein 4-beta-N-acetylglucosaminyltransferase C; GlcNAc-T IVc; gnT-IVc; GNTIVH mannosyl (alpha-13-)-glycoprotein beta-14-N-acetylglucosaminyltransferase isozyme C (putative) UDP-N-acetylglucosamine:a-13-D-mannoside beta-14-N-acetylglucosaminyltransferase IV; hGnT-IV-H; mannosyl (alpha-1,3-)-glycoprotein beta-1,4-N-acetylglucosaminyltransferase, isozyme C (putative); MGAT4 family, member C; N-acetylglucosaminyltransferase IV homolog; N-acetylglucosaminyltransferase IVc; N-glycosyl-oligosaccharide-glycoprotein N-acetylglucosaminyltransferase IVc; UDP-N-acetylglucosamine: alpha-1,3-D-mannoside beta-1,4-N-acetylglucosaminyltransferase IVc; UDP-N-acetylglucosamine:a-1,3-D-mannoside beta-1,4-N-acetylglucosaminyltransferase IV; UDP-N-acetylglucosamine:a-1,3-D-mannoside beta-1,4-N-acetylglucosaminyltransferase IV-homolog
Gene Aliases: GNTIVH; HGNT-IV-H; MGAT4C
UniProt ID: (Human) Q9UBM8
Entrez Gene ID: (Human) 25834
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.