Novus Biologicals
Manufacturer Code:NBP297462JF646
Catalog # NB043847
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Produced in rabbits immunized with E. coli-derived Human MGAT4A fragment. |
Conjugate | Janelia Fluor 646 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Alpha-1,3-mannosyl-glycoprotein 4-beta-N-acetylglucosaminyltransferase A; Alpha-1,3-mannosyl-glycoprotein 4-beta-N-acetylglucosaminyltransferase A soluble form; alpha-1,3-mannosyl-glycoprotein beta-1,4-N-acetylglucosaminyltransferase; alpha-13-mannosyl-glycoprotein 4-beta-N-acetylglucosaminyltransferase A alpha-13-mannosyl-glycoprotein beta-14-N-acetylglucosaminyltransferase EC 2.4.1.145 GlcNAc-T IVa GNT-IV GNT-IVA isoenzyme A isozyme A mannosyl (alpha-13-)-glycoprotein beta-14-N-acetylglucosaminyltransferase N-acetylglucosaminyltransferase IVa N-glycosyl-oligosaccharide-glycoprotein N-acetylglucosaminyltransferase IVa UDP-GlcNAc:a-13-D-mannoside b-14-acetylglucosaminyltransferase IV UDP-N-acetylglucosamine: alpha-13-D-mannosidebeta-14-N-acetylglucosaminyltransferase IVa UDP-N-acetylglucosamine:alpha13-d-mannosidebeta14-N-acetylglucosaminyltransferase; GlcNAc-T IVa; mannosyl (alpha-1,3-)-glycoprotein beta-1,4-N-acetylglucosaminyltransferase, isoenzyme A; N-acetylglucosaminyltransferase IVa; N-glycosyl-oligosaccharide-glycoprotein N-acetylglucosaminyltransferase IVa; UDP-GlcNAc:a-1,3-D-mannoside b-1,4-acetylglucosaminyltransferase IV; UDP-N-acetylglucosamine: alpha-1,3-D-mannoside beta-1,4-N-acetylglucosaminyltransferase IVa; UDP-N-acetylglucosamine:alpha1,3-d-mannoside beta1,4-N-acetylglucosaminyltransferase
Gene Aliases: GnT-4a; GNT-IV; GNT-IVA; MGAT4A
UniProt ID: (Human) Q9UM21
Entrez Gene ID: (Human) 11320
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.