Invitrogen
Manufacturer Code:PA597186
Catalog # PIPA597186
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit fusion protein containing a sequence corresponding to amino acids 178-447 of human MGAT2. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Alpha-1,6-mannosyl-glycoprotein 2-beta-N-acetylglucosaminyltransferase; Beta-1,2-N-acetylglucosaminyltransferase II; beta-12-N-acetylglucosaminyltransferase II; GlcNAc-T II; GNT-II; Mannoside acetylglucosaminyltransferase 2; N-glycosyl-oligosaccharide-glycoprotein N-acetylglucosaminyltransferase II; UDP-N-acetylglucosamine:alpha-6-D-mannoside beta-1,2-N-acetylglucosaminyltransferase II
Gene Aliases: AA407964; CDG2A; CDGS2; GLCNACTII; GNT-II; GNT2; MGAT2
UniProt ID: (Human) Q10469, (Mouse) Q921V5, (Rat) Q09326
Entrez Gene ID: (Human) 4247, (Mouse) 217664, (Rat) 94273
Molecular Function: glycosyltransferase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.