Invitrogen
Manufacturer Code:PA542625
Catalog # PIPA542625
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the middle region of human MGAT2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Alpha-1,6-mannosyl-glycoprotein 2-beta-N-acetylglucosaminyltransferase; Alpha-1,6-mannosyl-glycoprotein 2-beta-N-acetylglucosaminyltransferase-like protein; Beta-1,2-N-acetylglucosaminyltransferase II; beta-12-N-acetylglucosaminyltransferase II; GlcNAc-T II; GNT-II; Mannoside acetylglucosaminyltransferase 2; Mannosyl (alpha-1 6-)-glycoprotein beta-1 2-N-acetylglucosaminyltransferase CDGS2 GLCNACTII GNT-II GNT2 CDG2A; mannosyl (alpha-1,6-)-glycoprotein beta-1,2-N-acetylglucosaminyltransferase; N-glycosyl-oligosaccharide-glycoprotein N-acetylglucosaminyltransferase II; UDP-N-acetylglucosamine:alpha-6-D-mannoside beta-1,2-N-acetylglucosaminyltransferase II
Gene Aliases: AA407964; CDG2A; CDGS2; CH73-301C19.1; GLCNACTII; GNT-II; GNT2; MGAT2; zgc:162268
UniProt ID: (Human) Q10469, (Mouse) Q921V5, (Rat) Q09326
Entrez Gene ID: (Human) 4247, (Dog) 480312, (Rabbit) 100348676, (Horse) 100065740, (Mouse) 217664, (Guinea pig) 100724305, (Rat) 94273, (Zebrafish) 100000478
Molecular Function:
glycosyltransferase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.