Invitrogen
Manufacturer Code:PA530224
Catalog # PIPA530224
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit fragment corresponding to a region within amino acids 1 and 227 of Human MGAT2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Alpha-1,6-mannosyl-glycoprotein 2-beta-N-acetylglucosaminyltransferase; Beta-1,2-N-acetylglucosaminyltransferase II; CDG2A CDGS2 GLCNACTII GNT-II GNT2 Beta-12-N-acetylglucosaminyltransferase II N-glycosyl-oligosaccharide-glycoprotein N-acetylglucosaminyltransferase II UDP-N-acetylglucosamine:alpha-6-D-mannoside beta-12-N-acetylglucosaminyltransferase II alpha-16-mannosyl-glycoprotein 2-beta-N-acetylg; GlcNAc-T II; GNT-II; Mannoside acetylglucosaminyltransferase 2; N-glycosyl-oligosaccharide-glycoprotein N-acetylglucosaminyltransferase II; UDP-N-acetylglucosamine:alpha-6-D-mannoside beta-1,2-N-acetylglucosaminyltransferase II
Gene Aliases: CDG2A; CDGS2; GLCNACTII; GNT-II; GNT2; MGAT2
UniProt ID: (Human) Q10469
Entrez Gene ID: (Human) 4247
Molecular Function:
glycosyltransferase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.