Novus Biologicals
Manufacturer Code:NBP169335
Catalog # NBP169335
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptides corresponding to MGAT2(mannosyl (alpha-16-)-glycoprotein beta-12-N-acetylglucosaminyltransferase) The peptide sequence was selected from the middle region of MGAT2. Peptide sequence PKNAALKLGCINAEYPDSFGHYREAKFSQTKHHWWW |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Alpha-1,6-mannosyl-glycoprotein 2-beta-N-acetylglucosaminyltransferase; alpha-16-mannosyl-glycoprotein 2-beta-N-acetylglucosaminyltransferase Beta-12-N-acetylglucosaminyltransferase II CDG2A EC 2.4.1.143 GlcNAc-T II GLCNACTII GNT2 GNT-IICDGS2 Mannoside acetylglucosaminyltransferase 2 mannosyl (alpha-16-)-glycoprotein beta-12-N-acetylglucosaminyltransferase N-glycosyl-oligosaccharide-glycoprotein N-acetylglucosaminyltransferase II UDP-N-acetylglucosamine:alpha-6-D-mannosidebeta-12-N-acetylglucosaminyltransferase II; Beta-1,2-N-acetylglucosaminyltransferase II; GlcNAc-T II; GNT-II; Mannoside acetylglucosaminyltransferase 2; N-glycosyl-oligosaccharide-glycoprotein N-acetylglucosaminyltransferase II; UDP-N-acetylglucosamine:alpha-6-D-mannoside beta-1,2-N-acetylglucosaminyltransferase II
Gene Aliases: CDG2A; CDGS2; GLCNACTII; GNT-II; GNT2; MGAT2
UniProt ID: (Human) Q10469
Entrez Gene ID: (Human) 4247
Molecular Function:
glycosyltransferase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.