Invitrogen
Manufacturer Code:PA512147
Catalog # PIPA512147
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | KLH conjugated synthetic peptide between 16-46 amino acids from the N-terminal region of human MGAT1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Alpha-1,3-mannosyl-glycoprotein 2-beta-N-acetylglucosaminyltransferase; Alpha-13-mannosyl-glycoprotein 2-beta-N-acetylglucosaminyltransferase N-glycosyl-oligosaccharide-glycoprotein N-acetylglucosaminyltransferase I GNT-I GlcNAc-T I; glcNAc-T I; GNT-I; N-glycosyl-oligosaccharide-glycoprotein N-acetylglucosaminyltransferase I
Gene Aliases: GGNT1; GLCNAC-TI; GLCT1; GLYT1; GNT-1; GNT-I; Gnt1; GnTI; MGAT; Mgat-1; MGAT1
UniProt ID: (Human) P26572, (Mouse) P27808
Entrez Gene ID: (Human) 4245, (Mouse) 17308
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.