Novus Biologicals
Manufacturer Code:NBP30474120UL
Catalog # NB096086
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 156-445 of human MGAT1 (NP_001108092.1). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Alpha-1,3-mannosyl-glycoprotein 2-beta-N-acetylglucosaminyltransferase; alpha-13-mannosyl-glycoprotein 2-beta-N-acetylglucosaminyltransferase EC 2.4.1.101 GGNT1 GlcNAc-T I GLCT1 GLYT1GLCNAC-TI GNT-1 GNT-I mannosyl (alpha-13-)-glycoprotein beta-12-N-acetylglucosaminyltransferase MGAT N-glycosyl-oligosaccharide-glycoprotein N-acetylglucosaminyltransferase I; glcNAc-T I; GNT-I; N-glycosyl-oligosaccharide-glycoprotein N-acetylglucosaminyltransferase I
Gene Aliases: GGNT1; GLCNAC-TI; GLCT1; GLYT1; GNT-1; GNT-I; GnTI; MGAT; MGAT1
UniProt ID: (Human) P26572
Entrez Gene ID: (Human) 4245
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.