OriGene
Manufacturer Code:CF811799
Catalog # 50-167-0853
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI3D9 |
Immunogen | Full length human recombit protein of human MGARP produced in E.coli. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: C4orf49 CESP-1 HUMMR OSAP; CESP-1; Corneal endothelium-specific protein 1; Hypoxia up-regulated mitochondrial movement regulator protein; Mitochondria-localized glutamic acid-rich protein; Ovary-specific acidic protein; Protein MGARP
Gene Aliases: C4orf49; CESP-1; CESP1; GS3582; HUMMR; MGARP; OSAP
UniProt ID: (Human) Q8TDB4
Entrez Gene ID: (Human) 84709
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.