Abnova
Manufacturer Code:PAB27460
Catalog # 89-128-971
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (PFA fixed) (IHC (PFX)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant protein corresponding to amino acids of human MGAM. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C. For long term storage store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against recombinant MGAM.
For Research Use Only
Protein Aliases: Alpha-glucosidase; brush border hydrolase; Glucan 1,4-alpha-glucosidase; Glucoamylase; Maltase; maltase-glucoamylase (alpha-glucosidase); Maltase-glucoamylase, intestinal; MG; MGA
Gene Aliases: MG; MGA; MGAM; MGAML
UniProt ID: (Human) O43451
Entrez Gene ID: (Human) 8972
Molecular Function: glucosidase hydrolase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.